Learn More
Aviva Systems Biology Corp MNS1 Antibody - N-terminal region (ARP57223_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57223_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MNS1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human MNS1. Alternate names: FLJ11222, FLJ26051ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-282