Learn More
Enzo Life Sciences MnSOD (mouse), (recombinant) (His-tag) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISOD200D
Encompass_Preferred
Alternative name: Superoxide Dismutase, SOD2. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~25kDa. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 1.5 % glycerol and 80mM sodium chloride. Source: Produced in E. coli. Mouse Mn SOD is fused at the N-terminus to a His-tag. Long Term Storage: -80°C. Applications: WB Activity assay.
Catalog No. 50-200-7409