Learn More
Enzo Life Sciences MnSOD polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISOD111F
Encompass_Preferred
Host: Rabbit. Immunogen: Native rat Mn SOD. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Drosophila, Dog, Clam, Chicken, Bovine. Applications: ELISA, Flow Cytometry, IHC (PS), IP, WB. Purity: Immunoaffinity purified. Formulation: Liquid. In sodium phosphate buffer, pH 7.0, containing 15mg/ml BSA, 0.09% sodium azide, and 50% glycerol. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7408