Learn More
McMaster-Carr Modular DIN-Rail Mount Terminal Block
Supplier:  McMaster-Carr 7641K71
Encompass
Modular DIN-Rail Mount Terminal Block, with 2 Circuits, 300V AC/300V DC, 6mm Wide
Catalog No. NC1457114