Learn More
ATCC MNT-1; Melanoma; Human (Homo sapiens)
Catalog No. 502383512
Encompass
MNT-1 is a cell line exhibiting fibroblast morphology that was isolated from the lymph node of a patient with melanoma. This cell line is useful for studies of melanosome properties, melanosome biogenesis formation, influences on pigmentation in humans, and membrane trafficking.
Catalog No. 50-238-3512 Supplier ATCC Supplier No. CRL3450
MNT-1 is a cell line exhibiting fibroblast morphology that was isolated from the lymph node of a patient with melanoma. This cell line is useful for studies of melanosome properties, melanosome biogenesis formation, influences on pigmentation in humans, and membrane trafficking.