Learn More
Enzo Life Sciences MnTBAP chloride (10mg)
SDP
Catalog No. 502009177
Encompass_Preferred
Cell permeable superoxide dismutase (SOD) mimetic. Potent inhibitor of peroxynitrite-induced oxidative reactions (peroxynitrite scavenger), but not a scavenger of nitric oxide (NO). Inhibits neuronal apoptosis. Purity: ≥95% (TLC). Appearance: Black crystalline powder. Solubility: Dissolve at 0.1M in aqueous base, then add buffer to lower pH to pH 7.0 or higher as needed. Long Term Storage: -20°C.
Catalog No. 50-200-9177 Supplier Enzo Life Sciences Supplier No. ALX430069M010
Cell permeable superoxide dismutase (SOD) mimetic. Potent inhibitor of peroxynitrite-induced oxidative reactions (peroxynitrite scavenger), but not a scavenger of nitric oxide (NO). Inhibits neuronal apoptosis. Purity: ≥95% (TLC). Appearance: Black crystalline powder. Solubility: Dissolve at 0.1M in aqueous base, then add buffer to lower pH to pH 7.0 or higher as needed. Long Term Storage: -20°C.