Learn More
AdipoGen MnTBAP chloride
SDP
Catalog No. 502562362
Encompass_Preferred
Chemical. CAS 55266-18-7. Formula C48H28MnN4O8Cl. MW 879.2. Cell permeable superoxide dismutase SOD mimetic. Potent inhibitor of peroxynitrite-induced oxidative reactions peroxynitrite scavenger, but not a scavenger of nitric oxide NO. Neuronal apoptosis inhibitor. Protects T cells from superoxide generation, caspase-dependent DNA loss and cell death. Lipopolysaccharide-induced TNF-alpha production inhibitor, by prevention of intracellular ROS generation and subsequent inactivation of p38 MAPK and SAPK/JNK. .
Catalog No. 50-256-2362 Supplier AdipoGen Supplier No. AGCR10133M010
Chemical. CAS 55266-18-7. Formula C48H28MnN4O8Cl. MW 879.2. Cell permeable superoxide dismutase SOD mimetic. Potent inhibitor of peroxynitrite-induced oxidative reactions peroxynitrite scavenger, but not a scavenger of nitric oxide NO. Neuronal apoptosis inhibitor. Protects T cells from superoxide generation, caspase-dependent DNA loss and cell death. Lipopolysaccharide-induced TNF-alpha production inhibitor, by prevention of intracellular ROS generation and subsequent inactivation of p38 MAPK and SAPK/JNK. .