Learn More
Abcam MnTBAP chloride, Supero x ide dismutase (SOD) mimetic, 10MG
Catalog No. 502706566
Encompass_Preferred
MW 879.1 Da. Cell-permeable SOD mimetic. Potent peroxynitrite scavenger. Does not scavenge NO. Shows anti-inflammatory effects and reduces oxidative injury in vivo. Reduces ROS and shows a neuroprotective effect in spinal cord injury.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6566 Supplier Abcam Supplier No. AB14149610MG
MW 879.1 Da. Cell-permeable SOD mimetic. Potent peroxynitrite scavenger. Does not scavenge NO. Shows anti-inflammatory effects and reduces oxidative injury in vivo. Reduces ROS and shows a neuroprotective effect in spinal cord injury.
The product is subject to the following: Abcam Restricted Use Statement