Learn More
Apexbio Technology LLC MnTMPyP Pentachloride 125565-45-9 50mg
SDP
Catalog No. 5000090170
Encompass_Preferred
MnTMPyP Pentachloride (CAS 125565-45-9) is a manganese (III) porphyrin compound known for superoxide dismutase (SOD) mimetic activity It is designed to catalytically disproportionate superoxide anion radicals into molecular oxygen and hydrogen peroxide thereby modulating intracellular oxidative stress MnTMPyP Pentachloride exerts its biological activity primarily through catalytic scavenging of superoxide radicals In biomedical research MnTMPyP Pentachloride is frequently utilized to investigate cellular redox homeostasis oxidative injury pathways and mechanisms underlying free radical-related signaling events Based on these pharmacological properties MnTMPyP Pentachloride holds research potential in experimental models related to neurological cardiovascular and inflammatory diseases
Catalog No. 50-000-90170 Supplier Apexbio Technology LLC Supplier No. B603750
MnTMPyP Pentachloride (CAS 125565-45-9) is a manganese (III) porphyrin compound known for superoxide dismutase (SOD) mimetic activity It is designed to catalytically disproportionate superoxide anion radicals into molecular oxygen and hydrogen peroxide thereby modulating intracellular oxidative stress MnTMPyP Pentachloride exerts its biological activity primarily through catalytic scavenging of superoxide radicals In biomedical research MnTMPyP Pentachloride is frequently utilized to investigate cellular redox homeostasis oxidative injury pathways and mechanisms underlying free radical-related signaling events Based on these pharmacological properties MnTMPyP Pentachloride holds research potential in experimental models related to neurological cardiovascular and inflammatory diseases