Learn More
AdipoGen MnTMPyP pentachloride
SDP
Catalog No. 502562389
Encompass_Preferred
Chemical. CAS 100012-18-8. Formula C44H36MnN8Cl5. MW 909. Cell permeable superoxide dismutase SOD mimetic. Peroxynitrite scavenger. Displays a protective effect against H2O2 mediated injury. Neuroprotective. Oxidative stress inhibitor. Apoptosis inhibitor. Reduces inflammation.
Catalog No. 50-256-2389 Supplier AdipoGen Supplier No. AGCR10026M010
Chemical. CAS 100012-18-8. Formula C44H36MnN8Cl5. MW 909. Cell permeable superoxide dismutase SOD mimetic. Peroxynitrite scavenger. Displays a protective effect against H2O2 mediated injury. Neuroprotective. Oxidative stress inhibitor. Apoptosis inhibitor. Reduces inflammation.