Learn More
Ibt Bioservices IBT Mouse anti-Sudan Virus Glycoprotein mAb (2H5), Cat # 0202-029, Mouse, Monoclonal, Sudan Virus, Sudan glycoprotein, Boniface, Protein A/G, ELISA, Western Blot, Liquid, Storage: -80°C
SDP
Supplier:  Ibt Bioservices 0202029
Encompass_Preferred
Protein G purified mouse monoclonal antibody reactive to SUDV glycoprotein (GP). The antibody detects glycoprotein in virus-like particles (VLP) and SUDV glycoprotein without the transmembrane region (rGPdTM).Supplied in PBS in frozen format with no preservatives added. Store at +4°C for 2-3 weeks; -80°C for long-term
Catalog No. NC2604975