Learn More
Charles River Labs Cell Solutions Mobilized CD34+ Stem/Progenitor Cells, GCSF, 5 Million cells, Cryopreserved
Catalog No. 502256179
Encompass_Preferred
CD34+ stem cells are multipotent and can give rise to all cell types in blood. CD34+ cells are most known for its expression on hematopoietic progenitor cells found in bone marrow and cord blood. Donors are injected with G-CSF, which stimulates the bone marrow to produce a large number of hematopoietic and progenitor stem cells and mobilizes them into the peripheral blood stream. CD34+ cells are isolated using positive immunomagnetic cell separation procedures from a mobilized leukapheresis collection.
This item is not returnable. View return policy
Catalog No. 50-225-6179 Supplier Charles River Labs Cell Solutions Supplier No. M34CGCSF2
CD34+ stem cells are multipotent and can give rise to all cell types in blood. CD34+ cells are most known for its expression on hematopoietic progenitor cells found in bone marrow and cord blood. Donors are injected with G-CSF, which stimulates the bone marrow to produce a large number of hematopoietic and progenitor stem cells and mobilizes them into the peripheral blood stream. CD34+ cells are isolated using positive immunomagnetic cell separation procedures from a mobilized leukapheresis collection.