Learn More
Boca Scientific Inc Mobicol "Classic" with 2 different screw caps and bottom plug, without filters, 50/pack
Supplier:  Boca Scientific Inc M1002
Encompass
Mobicols are incredibly versatile 1ml columns that are an indispensable addition to every lab. They are compatible with sample sizes from small volumes to large volumes and can handle small or large wash buffer volumes as well. The M1002 Mobicol “Classic” is supplied with two screw caps (Luer-lock cap and screw cap) and a plug for the outlet. Small and large filters with different diameters and pore sizes (10 µm, 35 µm and 90 µm) are separately available. With these features, the Mobicol “Classic” can be used for a wide range of applications as both mini columns and spin columns.
Catalog No. NC9309391