Learn More
Boca Scientific Inc M105035F Mobicol "F" with fixed outlet plug, inserted large 35μm filter and 2 different screw caps, pack of 50
Supplier:  Boca Scientific Inc M105035F
Encompass
Mobicols are incredibly versatile 1ml columns that are an indispensable addition to every lab. The M105035F Mobicol “F” is supplied with a fixed outlet plug, inserted large 35 µm filter and 2 different screw caps. These columns are compatible with sample sizes from small volumes to large volumes and can handle small or large wash buffer volumes as well. With these features, the Mobicol “F” can be used for a variety of applications in affinity chromatography.
Catalog No. NC0180178