Learn More
Boca Scientific Inc Mobicol filters (small/lower), 35µm pore size, 50/pack
Supplier:  Boca Scientific Inc M513515
Encompass
This filter (small/lower) has a 35 µm pore size and is designed for use with Mobicol M1002, M1003, M1050 and M1053. 50 filters are included.
Catalog No. NC1712479