Learn More
Grainger Mobil Multipurpose Grease, NLGI Grade 2, Aluminum Complex, 13.7 oz.
Catalog No. NC0507053
Encompass_Preferred
Mobilgrease FM 222 is a multipurpose grease engineered for food processing equipment that operates in diverse environments, from oven to freezer temperatures. Formulated with an aluminum complex thickener and base oils, it provides lubrication across a wide range of speeds and loads, effectively handling wear and shock load conditions.
  • Excellent water resistance and rust inhibition for protection during frequent washdowns
  • Excellent thickener shear stability for consistent lubrication between relubrication cycles
  • NSF H1 registered for incidental food contact
  • Certified Kosher, Parve, and Halal
  • Manufactured in ISO 21469 registered facilities
Catalog No. NC0507053 Supplier Grainger Supplier No. 2RV34
Mobilgrease FM 222 is a multipurpose grease engineered for food processing equipment that operates in diverse environments, from oven to freezer temperatures. Formulated with an aluminum complex thickener and base oils, it provides lubrication across a wide range of speeds and loads, effectively handling wear and shock load conditions.
  • Excellent water resistance and rust inhibition for protection during frequent washdowns
  • Excellent thickener shear stability for consistent lubrication between relubrication cycles
  • NSF H1 registered for incidental food contact
  • Certified Kosher, Parve, and Halal
  • Manufactured in ISO 21469 registered facilities