Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Mobile Beta Storage Container
SCI_ED

Catalog No. s29142 Shop All Fisherbrand Products
Change view
Click to view available options
Height (Metric):
71 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
S29142 71 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S29142
This item is not returnable. View return policy

Suitable for beta protection and industrial mobile storage.

  • Made of 0.375 in. thick (0.95 cm) acrylic
  • Hinged lid stays open in two positions for safe, easy access
  • Four easy-glide casters
  • Dimensions: 20L x 20W x 28 in.H (50.8 x 50.8 x 71.1 cm)

Order Info

Additional mobile beta storage boxes sizes available. To order, contact your Fisher Scientific Customer Service Representative.

Specifications

Height (English) 28 in.
For Use With (Application) For beta protection and industrial mobile storage
Dimensions (L x W x H) 20 x 20 x 28 in. (50.8 x 50.8 x 71 cm)
Material Acrylic
Holds Carboys, linens, bench liners
Height (Metric) 71 cm
Includes Radiation hazard warning labels, 4 easy-glide casters, hinged lid
Length (English) 20 in.
Length (Metric) 50.8 cm
Thickness (English) 0.37 in.
Width (English) 20 in.
Product Type Beta storage container
Width (Metric) 50.8 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.