Learn More
Lakeside Manufacturing Inc Mobile Storage Cabinet with (5) Stainless Shelves
Catalog No. 502884350
Encompass_Preferred
Lakeside can design a custom wall of cabinetry that precisely fits your space and environment. The Lakeside 3679S Mobile Storage Cabinet has (5) stainless shelves. Durable construction with stainless steel body, top, bottom and door frames. 1/4" (6) thick clear tempered glass clearly displays contents. Counter weighted base for greater stability. Great for combination storage systems. Standard key lock. 4" (102) casters; 2 with brakes. Made in the USA.
Catalog No. 50-288-4350 Supplier Lakeside Manufacturing Inc Supplier No. 3679S
Add to Cart
Add to Cart
Request a Quote
Lakeside can design a custom wall of cabinetry that precisely fits your space and environment. The Lakeside 3679S Mobile Storage Cabinet has (5) stainless shelves. Durable construction with stainless steel body, top, bottom and door frames. 1/4" (6) thick clear tempered glass clearly displays contents. Counter weighted base for greater stability. Great for combination storage systems. Standard key lock. 4" (102) casters; 2 with brakes. Made in the USA.