Learn More
Description
- Provides stable support during use.
- Enables flexible workstation placement.
- Maintains safety and performance during operation.
- Features a Heavy-duty design with four casters for easy mobility.
- Constructed from durable steel.
- Compatible with the FS040 36 in. reveal ductless fume hood
- Allows easy movement around the laboratory while providing safe and stable support.
- Photo is for reference only, reveal ductless fume hood is sold separately.
Specifications
Specifications
| For Use With (Equipment) | 3 ft. Ductless Fume Hood |
| Product Type | Mobile Cart |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
