Learn More
Vetequip Inc V-10 Mobile Laboratory Animal Anesthesia System
SDP
Catalog No. NC9211433
Encompass_Preferred
Every "Lab Animal System" comes with an Accessory Kit containing a ten foot oxygen hose, flowmeter, oxygen flush assembly, vaporizer, breathing circuit, chamber, nosecones, waste gas evacuation tubing and two VaporGuard filters. One dial setting delivers a consistent and safe anesthetic level.All waste gases are scavenged for easy evacuation. Dual- & multi-patient circuits allow procedures to be handled in an assembly-line manner.Breathing circuits allow flat, prone or supine positions with head access. Positive sealing gasket on induction chambers minimize exposure to any waste gases.
Catalog No. NC9211433 Supplier Vetequip Inc Supplier No. 901807
 Restricted Item
Add to Cart
Add to Cart
Every "Lab Animal System" comes with an Accessory Kit containing a ten foot oxygen hose, flowmeter, oxygen flush assembly, vaporizer, breathing circuit, chamber, nosecones, waste gas evacuation tubing and two VaporGuard filters. One dial setting delivers a consistent and safe anesthetic level.All waste gases are scavenged for easy evacuation. Dual- & multi-patient circuits allow procedures to be handled in an assembly-line manner.Breathing circuits allow flat, prone or supine positions with head access. Positive sealing gasket on induction chambers minimize exposure to any waste gases.