Learn More
Microsolv Technology Corporation Mobile Phase Bottle 10L Sc
Catalog No. 5001452639
Encompass_Preferred
Bottle, Mobile Phase Reservoir, Safety, Coated Glass, Clear, 10L, Screw Top, GL45 Threads. MicroSolv Brand. 1/EA.
Catalog No. 50-014-52639 Supplier Microsolv Technology Corporation Supplier No. 5880210
Bottle, Mobile Phase Reservoir, Safety, Coated Glass, Clear, 10L, Screw Top, GL45 Threads. MicroSolv Brand. 1/EA.