Learn More
Microsolv Technology Corporation Mobile Phase Bottle 10L Uncoat
Catalog No. 5001452640
Encompass_Preferred
Bottle, Mobile Phase Reservoir, Glass, Clear, 10L, Screw Top, GL45 Threads. MicroSolv Brand. 1/EA.
Catalog No. 50-014-52640 Supplier Microsolv Technology Corporation Supplier No. 5880210X
Bottle, Mobile Phase Reservoir, Glass, Clear, 10L, Screw Top, GL45 Threads. MicroSolv Brand. 1/EA.