Learn More
Microsolv Technology Corporation Mobile Phase Bottle 1L S/T
Catalog No. 5001452635
Encompass_Preferred
Bottle, Mobile Phase Reservoir, Glass, Clear, 1L, Standard Taper Top, 29/32 Neck. MicroSolv Brand. 1/EA.
Catalog No. 50-014-52635 Supplier Microsolv Technology Corporation Supplier No. 5880101
Bottle, Mobile Phase Reservoir, Glass, Clear, 1L, Standard Taper Top, 29/32 Neck. MicroSolv Brand. 1/EA.