Learn More
Microsolv Technology Corporation Mobile Phase Bottle 1L S/T Co
Catalog No. 5001452636
Encompass_Preferred
Bottle, Mobile Phase Reservoir, Safety, Coated Glass, Clear, 1L, Standard Taper Top, 29/32 Neck. MicroSolv Brand. 1/EA.
Catalog No. 50-014-52636 Supplier Microsolv Technology Corporation Supplier No. 5880102
Bottle, Mobile Phase Reservoir, Safety, Coated Glass, Clear, 1L, Standard Taper Top, 29/32 Neck. MicroSolv Brand. 1/EA.