Learn More
Microsolv Technology Corporation Mobile Phase Bottle 1L Sc
Catalog No. 5001452637
Encompass_Preferred
Bottle, Mobile Phase Reservoir, Safety, Coated Glass, Clear, 1L, Screw Top, GL45 Threads. MicroSolv Brand. 1/EA.
Catalog No. 50-014-52637 Supplier Microsolv Technology Corporation Supplier No. 5880201
Bottle, Mobile Phase Reservoir, Safety, Coated Glass, Clear, 1L, Screw Top, GL45 Threads. MicroSolv Brand. 1/EA.