Learn More
Microsolv Technology Corporation Mobile Phase Bottle Red Cap
Catalog No. 5001452648
Encompass_Preferred
Bottle, Mobile Phase Reservoir, Safety, Coated Glass, Clear, 2L, Screw Top, GL45 Threads, Open Red Cap. MicroSolv Brand. 1/EA.
Catalog No. 50-014-52648 Supplier Microsolv Technology Corporation Supplier No. 5881905R
Bottle, Mobile Phase Reservoir, Safety, Coated Glass, Clear, 2L, Screw Top, GL45 Threads, Open Red Cap. MicroSolv Brand. 1/EA.