Learn More
Microsolv Technology Corporation Mobile Phase Filter, 10um, Stainless Steel, Tripod Tubing Connector. 1 EA.
Supplier:  Microsolv Technology Corporation 4822006
Encompass_Preferred
These Stainless Steel filters are manufactured to provide very high filter capacity and throughput. The 10um pore size will filter most undissolved particles from your solvents before they enter your HPLC pumps, columns and instrument but will produce less resistance to the vacuum and the in-flow of solvents compared to the 2um filter. The all PEEK Tripod Connector allows for the inlet tubing of the mobile phase delivery system to slip over it making it usable with 1/16" and 1/8" low pressure tubing. No ferrules or nuts are required. Install these filters directly in your mobile phase reservoir bottles.
Catalog No. 50-227-1514