Learn More
Sigma Aldrich Fine Chemicals Biosciences Supelco Mobile Phase Filtration Apparatus 2, 1 each
Catalog No. NC1754997
Encompass
Connects to aspiration line. Includes: 250 mL glass reservoir. S/T 34/45 funnel base. S/T 34/45 1000 mL flask and glass cap. Clamp. Stainless steel holder and screen. 10 PTFE gaskets. 50 Nylon 66 filters (47 mm, 0.45 um pores).
Catalog No. NC1754997 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 58062U
Connects to aspiration line. Includes: 250 mL glass reservoir. S/T 34/45 funnel base. S/T 34/45 1000 mL flask and glass cap. Clamp. Stainless steel holder and screen. 10 PTFE gaskets. 50 Nylon 66 filters (47 mm, 0.45 um pores).