Learn More
MilliporeSigma™ Supelco™ Mobile Phase Filtration Apparatus 2
Catalog No. 111008848 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Includes: 250 mL glass reservoir, S/T 34/45 funnel base, S/T 34/45 1000 mL flask and glass cap, Clamp, Stainless steel holder and screen, 10 PTFE gaskets and 50 Nylon 66 filters (47 mm, 0.45 um pores).
Catalog No.
11-100-8848 Includes: 250 mL glass reservoir, S/T 34/45 funnel base, S/T 34/45 1000 mL flask and glass cap, Clamp, Stainless steel holder and screen, 10 PTFE gaskets and 50 Nylon 66 filters (47 mm, 0.45 um pores).
1 options
Catalog No. 11-100-8848 Supplier MilliporeSigma™ Supelco™ Supplier No. 58062U
Add to Cart
Add to Cart
Connects to aspiration line
TRUSTED_SUSTAINABILITY
Specifications
Description Includes: 250 mL glass reservoir, S/T 34/45 funnel base, S/T 34/45 1000 mL flask and glass cap, Clamp, Stainless steel holder and screen, 10 PTFE gaskets and 50 Nylon 66 filters (47 mm, 0.45 μm pores).