Learn More
MilliporeSigma™ Supelco™ Mobile Phase Solvent Debubbler with Bracket
Catalog No. 111010638 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Application):
HPLC
Catalog No. For Use With (Application)
11-101-0638 HPLC
1 options
Catalog No. 11-101-0638 Supplier MilliporeSigma™ Supelco™ Supplier No. 58453
Gas bubbles in the mobile phase reduce pump output and cause check valves to malfunction. The mobile phase debubbler removes bubbles from the pump inlet line. Bubbles in the incoming mobile phase rise in the debubbler, displacing an equal volume of liquid from the debubbler reservoir to the pump. When necessary, simply remove the cap and refill the reservoir -- the gas-liquid interface is visible through the housing.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) HPLC
Description Compatible with 1.5 mm I.D. and 3.0 mm I.D. PTFE tubing used with Waters and other popular HPLC pumps. KEL-F, PTFE, and 316 stainless steel construction.