Learn More
Medicus Health Mobile Phlebotomy Draw Station with Work Surface, 8 Tube
SDP
Catalog No. NC2261985
Encompass
∙ Mobile Phlebotomy Draw Station with Work Surface is a portable """"workstation"""" that can travel anywhere ∙ Mobile Phlebotomy Draw Station with Work Surface enhances efficiency by keeping supplies in full view and close to your patient ∙ Ideal for holding safety devices, tourniquets, and draw accessories ∙ Great organization solution for any small area ∙ Spring securely clamps on chair arms, wheelchairs and bed rails ∙ 8 tube is designed to hold six 17mm tubes, four 13mm tubes, two 11mm tubes, alcohol wipes, and adhesive bandages ∙ Overall dimensions: 6""""W x 7""""L x 6""""H
This item is not returnable. View return policy
Catalog No. NC2261985 Supplier Medicus Health Supplier No. 7303M1
∙ Mobile Phlebotomy Draw Station with Work Surface is a portable """"workstation"""" that can travel anywhere ∙ Mobile Phlebotomy Draw Station with Work Surface enhances efficiency by keeping supplies in full view and close to your patient ∙ Ideal for holding safety devices, tourniquets, and draw accessories ∙ Great organization solution for any small area ∙ Spring securely clamps on chair arms, wheelchairs and bed rails ∙ 8 tube is designed to hold six 17mm tubes, four 13mm tubes, two 11mm tubes, alcohol wipes, and adhesive bandages ∙ Overall dimensions: 6""""W x 7""""L x 6""""H