Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hughes Mobile Self-Contained Hand Washing Station

Catalog No. 19111124 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (English):
30 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-111-124 30 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-111-124 Supplier Hughes Supplier No. HW40K
  • $14,837.00
    $12,196.65 / Each of 1

    Save $2,640.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

This self-contained, portable 30 gallon capacity hand washing station provides a reliable way for personnel to ensure they are able to manage an effective hygiene regime, whatever the location. The station delivers a controlled flow of water to perform approximately 150 hand washes.

This self-contained, portable 30 gallon capacity hand washing station provides a reliable way for personnel to ensure they are able to manage an effective hygiene regime, whatever the location. The station delivers a controlled flow of water to perform approximately 150 hand washes* from a full unit.

Hands-free, foot activation for ease of use and to minimize cross-contamination. Delivers a controlled flow of water to wash hands properly. Approximately 150 hand washes for a full unit (unit has a 30 gallon capacity). Simple to fill with a standard garden hose via the 1/2 inch quick release water inlet connection. Stainless-steel frame and large pneumatic tires aid portability making it easy to maneuver by one person. Soap and paper towel dispenser (contents not provided). Can act as a supplement to permanent installations. Suitable for indoor or outdoor use.

Please note:
Used water is not contained but drains to the base of the station and should be directed to flow away from the unit or to an appropriate drainage point to avoid a slip hazard.

*Based on a 20 second hand wash using water only when rinsing.

Specifications

Capacity (English) 30 gal.
Capacity (Metric) 114 L
Color Gray, White, Black, Blue
Depth (English) 44 in.
Depth (Metric) 1254 mm
Flow Rate 2 lpm at 1.2 Bar
For Use With (Application) Portable Hand Washing Station, COVID-19 Prevention
For Use With (Equipment) Portable, No additional Equipment needed
Height (English) 69 in.
Height (Metric) 1752 mm
Includes All in One unit
Material Steel, Plastic
Product Type Hand Washing Station
Sterility Non-sterile
Storage Requirements Operating Temp.: 41 to 95°F (5 to 35°C)
Weight (English) 176 lbs.
Weight (Metric) 80 kg
Width (English) 33 in.
Width (Metric) 839 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.