Learn More
Andwin Scientific MOBILE TUB LDPE 28GAL EA
SDP
Catalog No. 501305852
Encompass_Preferred
With a rugged caster frame, this rectangular mobile tub is able to store heavy solids or liquids of up to 106 liters. The tub and caster can easily be removed and cleaned. These products are approved by USDA and certified by NSF. A polyethylene cover is included.
Catalog No. 50-130-5852 Supplier Andwin Scientific Supplier No. 674540A
With a rugged caster frame, this rectangular mobile tub is able to store heavy solids or liquids of up to 106 liters. The tub and caster can easily be removed and cleaned. These products are approved by USDA and certified by NSF. A polyethylene cover is included.