Learn More
Lakeside Manufacturing Inc Mobile Wide Storage Cabinet with (5) Stainless Steel Shelves, Sloped Top
Catalog No. 502884424
Encompass_Preferred
Lakeside can design a custom wall of cabinetry that precisely fits your space and environment. The Lakeside ST-4879S Mobile Wide Storage Cabinet has (5) stainless steel shelves and a sloped top. Durable construction with stainless steel body, top, bottom and door frames. 1/4" (6) thick clear tempered glass clearly displays contents. Counter weighted base for greater stability. Great for combination storage systems. Standard key lock. 4" (102) casters; 2 with brakes. Optional sloping top prevents the accumulation of dust. Made in the USA.
Catalog No. 50-288-4424 Supplier Lakeside Manufacturing Inc Supplier No. ST4879S
Add to Cart
Add to Cart
Request a Quote
Lakeside can design a custom wall of cabinetry that precisely fits your space and environment. The Lakeside ST-4879S Mobile Wide Storage Cabinet has (5) stainless steel shelves and a sloped top. Durable construction with stainless steel body, top, bottom and door frames. 1/4" (6) thick clear tempered glass clearly displays contents. Counter weighted base for greater stability. Great for combination storage systems. Standard key lock. 4" (102) casters; 2 with brakes. Optional sloping top prevents the accumulation of dust. Made in the USA.