Learn More
Lakeside Manufacturing Inc Mobile Wide Storage Cabinet with (5) Stainless Steel Shelves
Catalog No. 502884457
Encompass_Preferred
Lakeside can design a custom wall of cabinetry that precisely fits your space and environment. The Lakeside 4879S Mobile Wide Storage Cabinet has (5) stainless steel shelves. Durable construction with stainless steel body, top, bottom and door frames. 1/4" (6) thick clear tempered glass clearly displays contents. Counter weighted base for greater stability. Great for combination storage systems. Standard key lock. 4" (102) casters; 2 with brakes. Made in the USA.
Catalog No. 50-288-4457 Supplier Lakeside Manufacturing Inc Supplier No. 4879S
Add to Cart
Add to Cart
Request a Quote
Lakeside can design a custom wall of cabinetry that precisely fits your space and environment. The Lakeside 4879S Mobile Wide Storage Cabinet has (5) stainless steel shelves. Durable construction with stainless steel body, top, bottom and door frames. 1/4" (6) thick clear tempered glass clearly displays contents. Counter weighted base for greater stability. Great for combination storage systems. Standard key lock. 4" (102) casters; 2 with brakes. Made in the USA.