Learn More
Lakeside Manufacturing Inc Mobile Wide Storage Cabinet with Stainless Steel Shelves, Glass Doors
Catalog No. 502884520
Encompass_Preferred
Lakeside can design a custom wall of cabinetry that precisely fits your space and environment. The Lakeside 4724G Mobile Wide Storage Cabinet has stainless steel shelves and glass doors. Durable construction with stainless steel body, top, bottom and door frames. 1/4" (6) thick clear tempered glass clearly displays contents. Counter weighted base for greater stability. Great for combination storage systems. Standard key lock. 4" (102) casters; 2 with brakes. Made in the USA.
Catalog No. 50-288-4520 Supplier Lakeside Manufacturing Inc Supplier No. 4724G
Add to Cart
Add to Cart
Request a Quote
Lakeside can design a custom wall of cabinetry that precisely fits your space and environment. The Lakeside 4724G Mobile Wide Storage Cabinet has stainless steel shelves and glass doors. Durable construction with stainless steel body, top, bottom and door frames. 1/4" (6) thick clear tempered glass clearly displays contents. Counter weighted base for greater stability. Great for combination storage systems. Standard key lock. 4" (102) casters; 2 with brakes. Made in the USA.