Learn More
Mascon Medical MobiNova®-D1 Dissociation Tubes, 25 pieces
SDP
Catalog No. NC3802108
Encompass_Preferred
Consumables used in conjunction with the MobiNova-D1. Used for preparing single-cell suspensions for downstream single-cell sequencing, cell sorting, and other experiments.
Catalog No. NC3802108 Supplier Mascon Medical Supplier No. MMDT25
Consumables used in conjunction with the MobiNova-D1. Used for preparing single-cell suspensions for downstream single-cell sequencing, cell sorting, and other experiments.