Learn More
Mascon Medical MobiSolution Multi Tissue Dissociation Kit, Mouse, 25rxn
SDP
Catalog No. NC3802110
Encompass_Preferred
Reagents used in conjunction with the MobiNova-D1. The function of the mouse multi tissue dissociation kit is to gently and efficiently prepare single-cell suspensions from various mouse tissues. Its performance has been validated in various mouse tissues, including liver, spleen, lung, kidney, intestine, etc.
Catalog No. NC3802110 Supplier Mascon Medical Supplier No. MMDRM25
Reagents used in conjunction with the MobiNova-D1. The function of the mouse multi tissue dissociation kit is to gently and efficiently prepare single-cell suspensions from various mouse tissues. Its performance has been validated in various mouse tissues, including liver, spleen, lung, kidney, intestine, etc.