Learn More
Mascon Medical MobiSolution Tumor Dissociation Kit,Human, 25rxn
SDP
Catalog No. NC3802109
Encompass_Preferred
Reagents used in conjunction with the MobiNova-D1.The tumor dissociation kit is developed for the dissociation of primary human tumors and xenografts. It has been optimized for a variety of tumor types, including breast cancer, pancreatic cancer, prostate cancer, colon cancer, lung adenocarcinoma, etc.
Catalog No. NC3802109 Supplier Mascon Medical Supplier No. MMDRH25
Reagents used in conjunction with the MobiNova-D1.The tumor dissociation kit is developed for the dissociation of primary human tumors and xenografts. It has been optimized for a variety of tumor types, including breast cancer, pancreatic cancer, prostate cancer, colon cancer, lung adenocarcinoma, etc.