Learn More
MilliporeSigma Single-use 3L stirred tank bioreactor (green), 2/pk
Catalog No. NC0285848
Encompass
A single-use, stirred tank bioreactor designed for the non-GMP cultivation of mammalian cell lines. Its rigid design includes a semi-hemispherical shaped polycarbonate vessel, a multi-port high-density polyethylene (HDPE) headplate, a scoping marine impeller, both open pipe and sintered spargers, weldable C-Flex tubing, and all necessary gas inlet and outlet filters.
Catalog No. NC0285848 Supplier MilliporeSigma Supplier No. CR0003L200
A single-use, stirred tank bioreactor designed for the non-GMP cultivation of mammalian cell lines. Its rigid design includes a semi-hemispherical shaped polycarbonate vessel, a multi-port high-density polyethylene (HDPE) headplate, a scoping marine impeller, both open pipe and sintered spargers, weldable C-Flex tubing, and all necessary gas inlet and outlet filters.