Learn More
Aviva Systems Biology Corp MOBKL2A Antibody - middle region (ARP52542_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52542_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MOBKL2A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human MOBKL2A. Alternate names: moblak, MOB1C, MOBKL2AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-728