Learn More
AllCells LLC Mobilized Peripheral Blood- Cryopreserved- Neupogen- CD34+ Stem/Progenitor Cells- Positively Selected- 5M
Catalog No. NC2742204
Encompass
CD34+ Stem/Progenitor cells are positively selected from Regimen F mobilized peripheral blood leukapheresis using immunomagnetic CD34+ microbeads. CD34+ cells are shipped in a 2mL cryovial. Purity is determined prior to cryopreservation and is ≥ 90%. Viability is ≥ 80% post-thaw. 5 Million Cells per vialAllCells is the industry leader in supplying mobilized Leukopaks with the shortest lead time on the market. We offer three different mobilization regimens: G-CSF (Neupogen)- plerixafor (Mozobil)- and a combination of the two drugs.
Catalog No. NC2742204 Supplier AllCells LLC Supplier No. MLPREGFCRCD34PS5M
CD34+ Stem/Progenitor cells are positively selected from Regimen F mobilized peripheral blood leukapheresis using immunomagnetic CD34+ microbeads. CD34+ cells are shipped in a 2mL cryovial. Purity is determined prior to cryopreservation and is ≥ 90%. Viability is ≥ 80% post-thaw. 5 Million Cells per vialAllCells is the industry leader in supplying mobilized Leukopaks with the shortest lead time on the market. We offer three different mobilization regimens: G-CSF (Neupogen)- plerixafor (Mozobil)- and a combination of the two drugs.