Learn More
Cayman Chemical MobocertInIb 10mg
SDP
Catalog No. 502991066
Encompass_Preferred
An inhibitor of mutant EGFR and HER2 receptors; selective for HER2V659E, HER2G660D, HER2G309E, and HER2S310F over wild-type EGFR in Ba/F3 cells
Catalog No. 50-299-1066 Supplier Cayman Chemical Supplier No. 3152210 MG
An inhibitor of mutant EGFR and HER2 receptors; selective for HER2V659E, HER2G660D, HER2G309E, and HER2S310F over wild-type EGFR in Ba/F3 cells