Learn More
AllCells LLC Mobilized Peripheral Blood- Cryopreserved- Neupogen and Mozobil- CD34+ Positively Selected 1M
Catalog No. NC2742211
Encompass
CD34+ Stem/Progenitor cells are positively selected from Regimen H mobilized peripheral blood leukapheresis using immunomagnetic CD34+ microbeads. CD34+ cells are shipped in a 2mL cryovial. Purity is determined prior to cryopreservation and is ≥ 90%. Viability is ≥ 80% post-thaw. 1 Million Cells per vial.AllCells is the industry leader in supplying mobilized Leukopaks with the shortest lead time on the market. We offer three different mobilization regimens: G-CSF (Neupogen)- plerixafor (Mozobil)- and a combination of the two drugs.
Catalog No. NC2742211 Supplier AllCells LLC Supplier No. MLPREGHCRCD34PS1M
CD34+ Stem/Progenitor cells are positively selected from Regimen H mobilized peripheral blood leukapheresis using immunomagnetic CD34+ microbeads. CD34+ cells are shipped in a 2mL cryovial. Purity is determined prior to cryopreservation and is ≥ 90%. Viability is ≥ 80% post-thaw. 1 Million Cells per vial.AllCells is the industry leader in supplying mobilized Leukopaks with the shortest lead time on the market. We offer three different mobilization regimens: G-CSF (Neupogen)- plerixafor (Mozobil)- and a combination of the two drugs.