Learn More
Biocare Medical, LLC MOC-31 Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC CM403C
Encompass_Preferred
MOC-31 antibody, also known as Epithelial Specific Antigen/Ep-CAM, recognizes an epithelial-associated, glycoprotein located on the cell membrane surface and in the cytoplasm of virtually all epithelial cells. It is not present in most squamous epithelia, hepatocytes, renal proximal tubular cells, gastric parietal cells and myoepithelial cells. MOC-31 may be used in a panel of antibodies as a negative marker for mesothelioma, or lung adenocarcinoma. Studies have shown that MOC-31 is useful in differentiating tumors of unknown origin in liver cancers and distinguishing cholangiocarcinoma from hepatocellular carcinomas.
Catalog No. 50-826-88