Learn More
Cayman Chemical Mocetinostat, Purity: ≥98%, CAS: 726169-73-9, 10 mg
SDP
Catalog No. 502979810
Encompass_Preferred
An orally available HDAC inhibitor that selectively targets HDAC1 and 2 (IC50s = 0.15 and 0.29 µM, respectively), less potently inhibits HDAC3 and 11 (IC50s = 1.66 and 0.59 µM, respectively); induces hyperacetylation of histones, causes cell cycle blockade, and induces apoptosis in cancer cells in vitro
Catalog No. 50-297-9810 Supplier Cayman Chemical Supplier No. 1828710 MG
An orally available HDAC inhibitor that selectively targets HDAC1 and 2 (IC50s = 0.15 and 0.29 µM, respectively), less potently inhibits HDAC3 and 11 (IC50s = 1.66 and 0.59 µM, respectively); induces hyperacetylation of histones, causes cell cycle blockade, and induces apoptosis in cancer cells in vitro