Learn More
Medchemexpress LLC HY-12164 100mg , Mocetinostat MGCD0103 CAS:726169-73-9 Purity:98%
SDP
Catalog No. 501492731
Encompass_Preferred
Medchemexpress, HY-12164 100mg Mocetinostat MGCD0103 CAS:726169-73-9 Formula:C23H20N6O IC50: 0.15 μM (HDAC1), 0.29 μM (HDAC2), 1.66 μM (HDAC3), 0.59 μM (HDAC11) Purity:98% Mocetinostat is a potent, orally active and isotype-selective HDAC (Class I/IV) inhibitor with IC 50 s of 0.15, 0.29, 1.66 and 0.59 μM for HDAC1 , HDAC2 , HDAC3 and HDAC11 , respectively; Mocetinostat shows no inhibition on HDAC4, HDAC5, HDAC6, HDAC7, or HDAC8. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2731 Supplier Medchemexpress LLC Supplier No. HY12164100MG
Medchemexpress, HY-12164 100mg Mocetinostat MGCD0103 CAS:726169-73-9 Formula:C23H20N6O IC50: 0.15 μM (HDAC1), 0.29 μM (HDAC2), 1.66 μM (HDAC3), 0.59 μM (HDAC11) Purity:98% Mocetinostat is a potent, orally active and isotype-selective HDAC (Class I/IV) inhibitor with IC 50 s of 0.15, 0.29, 1.66 and 0.59 μM for HDAC1 , HDAC2 , HDAC3 and HDAC11 , respectively; Mocetinostat shows no inhibition on HDAC4, HDAC5, HDAC6, HDAC7, or HDAC8. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.