Learn More
Medchemexpress LLC HY-12164 50mg , Mocetinostat CAS:726169-73-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1216450MG
Encompass_Preferred
HY-12164 50mg Mocetinostat CAS: 726169-73-9 Mocetinostat (MGCD0103) is a potent, orally active and isotype-selective HDAC (Class I/IV) inhibitor with IC50s of 0.15, 0.29, 1.66 and 0.59 μM for HDAC1, HDAC2, HDAC3 and HDAC11, respectively. Mocetinostat shows no inhibition on HDAC4, HDAC5, HDAC6, HDAC7, or HDAC8. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0610