Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mocetinostat | 726169-73-9 | >98.0% | 200 MG
SDP

Catalog No. 5000234791
Encompass_Preferred

Mocetinostat is a benzamide histone deacetylase (HDAC) inhibitor selective for class I/IV HDACs, used as a research compound in epigenetics and oncology studies. Typical potency: HDAC1 0.15 μM; HDAC2 0.29 μM; HDAC3 1.66 μM; HDAC11 0.59 μM. For research use only.

  • Selective class I/IV HDAC inhibitor.
  • Used in epigenetics and cancer research.
  • Typical purity >98% (supplier dependent).
  • Molecular formula C23H20N6O; molecular weight ≈396.4 g/mol.
  • Supplied as a 200 mg research pack.

Catalog No. 50-002-34791 Supplier Medchemexpress LLC Supplier No. HY12164200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mocetinostat is a benzamide histone deacetylase (HDAC) inhibitor selective for class I/IV HDACs, used as a research compound in epigenetics and oncology studies. Typical potency: HDAC1 0.15 μM; HDAC2 0.29 μM; HDAC3 1.66 μM; HDAC11 0.59 μM. For research use only.

  • Selective class I/IV HDAC inhibitor.
  • Used in epigenetics and cancer research.
  • Typical purity >98% (supplier dependent).
  • Molecular formula C23H20N6O; molecular weight ≈396.4 g/mol.
  • Supplied as a 200 mg research pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.