Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Moclobemide | 71320-77-9 | 99.1% | 50 MG
SDP

Catalog No. 501874119
Encompass_Preferred

Moclobemide is a brain-penetrant and reversible monoamine oxidase (MAO-A) inhibitor with an IC50 of 6.061 μM for hMAO-A. It up-regulates the proliferation of hippocampal progenitor cells in chronically stressed mice.

  • Brain-penetrant and reversible MAO-A inhibitor
  • Up-regulates proliferation of hippocampal progenitor cells
  • High purity of 99.1%
  • Soluble in DMSO at 100 mg/mL
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • Storage in solvent: -80°C for 2 years or -20°C for 1 year

Catalog No. 50-187-4119 Supplier Medchemexpress LLC Supplier No. HYB053450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Moclobemide is a brain-penetrant and reversible monoamine oxidase (MAO-A) inhibitor with an IC50 of 6.061 μM for hMAO-A. It up-regulates the proliferation of hippocampal progenitor cells in chronically stressed mice.

  • Brain-penetrant and reversible MAO-A inhibitor
  • Up-regulates proliferation of hippocampal progenitor cells
  • High purity of 99.1%
  • Soluble in DMSO at 100 mg/mL
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • Storage in solvent: -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.